Invitrogen
Manufacturer Code:PA5115909
Catalog # PIPA5115909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC18A1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4832416I10Rik CGAT Chromaffin granule amine transporter Slc18a1 solute carrier family 18 (vesicular monoamine transporter) member 1 solute carrier family 18 (vesicular monoamine) member 1 solute carrier family 18 member 1 solute carrier family 18 member A1 solute carrier family 18 member 1 solute carrier family18 (vesicular monoamine) member 1 (chromaffin granule amine transporter) VAT1 vesicular amine transporter 1 vesicular monoamine transporter 1 Vmat1 VMAT-1; Chromaffin granule amine transporter; solute carrier family 18 (vesicular monoamine transporter), member 1; solute carrier family 18 (vesicular monoamine), member 1; Solute carrier family 18 member 1; solute carrier family 18, member 1; solute carrier family18 (vesicular monoamine) member 1 (chromaffin granule amine transporter); VAT1; Vesicular amine transporter 1; vesicular monoamine transporter 1; VMAT1
Gene Aliases: 4832416I10Rik; CGAT; SLC18A1; VAT1; VMAT-1; VMAT1
UniProt ID: (Human) P54219, (Mouse) Q8R090, (Rat) Q01818
Entrez Gene ID: (Human) 6570, (Mouse) 110877, (Rat) 25693
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.