Invitrogen
Manufacturer Code:PA5115908
Catalog # PIPA5115908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC16A8 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 3; MCT 3 MCT3 monocarboxylate transporter Monocarboxylate transporter 3 preferentially expressed in RPE cells and choroid plexus epithelium proton-coupled monocarboxylate transporter 3 REMP Slc16a8 solute carrier 16 (monocarboxylic acid transporters) member 8 solute carrier family 16 (monocarboxylate transporter) member 8 solute carrier family 16 (monocarboxylic acid transporters) member 8 solute carrier family 16 member 8 solute carrier family 16 member 8 (monocarboxylic acid transporter 3); Monocarboxylate transporter 3; preferentially expressed in RPE cells and choroid plexus epithelium; Proton-coupled monocarboxylate transporter 3; solute carrier 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 (monocarboxylate transporter), member 8; solute carrier family 16 (monocarboxylic acid transporters), member 8; Solute carrier family 16 member 8; solute carrier family 16, member 8 (monocarboxylic acid transporter 3)
Gene Aliases: MCT3; REMP; SLC16A8
UniProt ID: (Human) O95907, (Mouse) O35308
Entrez Gene ID: (Human) 23539, (Mouse) 57274, (Rat) 65200
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.