Abnova
Manufacturer Code:PAB25666
Catalog # 89-127-285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids at internal region of human SHROOM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SHROOM3.
For Research Use Only
Protein Aliases: APXL3; F-actin-binding protein; hShrmL; KIAA1481; MSTP013; Protein Shroom3; SHRM; ShrmL; Shroom-related protein
Gene Aliases: APXL3; KIAA1481; MSTP013; SHRM; SHRML; SHROOM3
UniProt ID: (Human) Q8TF72
Entrez Gene ID: (Human) 57619
Molecular Function: actin family cytoskeletal protein cytoskeletal protein ion channel ligand-gated ion channel non-motor actin binding protein receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.