Invitrogen
Manufacturer Code:PA543467
Catalog # PIPA543467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SGPP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSPP1; hSPPase1; Sphingosine-1-phosphatase 1; Sphingosine-1-phosphate phosphatase 1; Sphingosine-1-phosphate phosphatase 1 SPPase1; sphingosine-1-phosphate phosphohydrolase; Sphingosine-1-phosphate phosphohydrolase 1; SPP; SPP-1; spp1; SPPase1
Gene Aliases: AI463453; SGPP1; SPP; SPP1; SPPase1; Spph1
UniProt ID: (Human) Q9BX95, (Mouse) Q9JI99
Entrez Gene ID: (Human) 81537, (Horse) 100061850, (Bovine) 516881, (Rabbit) 108178643, (Dog) 490728, (Guinea pig) 100715609, (Rat) 81536, (Mouse) 81535, (Zebrafish) 100003745
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.