R&D Systems
Manufacturer Code:AF5535
Catalog # AF5535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human SGPL1 Lys342-His568 Accession # O95470 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 4.1.2.27 FLJ13811 hSPL KIAA1252 S1PL Sphingosine-1-phosphate aldolase sphingosine-1-phosphate lyase 1 SPL SPLSPL 1 SP-lyase SP-lyase 1; hSPL; S1PL; SP-lyase 1; Sphingosine-1-phosphate aldolase; Sphingosine-1-phosphate lyase 1; SPL 1
Gene Aliases: KIAA1252; S1PL; SGPL1; SPL
UniProt ID: (Human) O95470
Entrez Gene ID: (Human) 8879
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.