Invitrogen
Manufacturer Code:PA112722
Catalog # 01-674-851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to sphingosine 1-phosphate lyase 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI428538 D10Xrf456 Spl; hSPL; mSPL; S1PL; SP-lyase 1; sphingosine phosphate lyase 1; Sphingosine-1-phosphate aldolase; Sphingosine-1-phosphate lyase 1; SPL 1
Gene Aliases: AI428538; D10Xrf456; KIAA1252; S1PL; SGPL1; Spgl1; SPL
UniProt ID: (Human) O95470, (Mouse) Q8R0X7, (Rat) Q8CHN6
Entrez Gene ID: (Human) 8879, (Bovine) 522515, (Mouse) 20397, (Rat) 286896
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.