Invitrogen
Manufacturer Code:PA5102733
Catalog # PIPA5102733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SGCE corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dystonia 11 myoclonic DYT11 EMBL:AAO37579.1 ECO:0000312 EMBL:AAO37579.1} Epsilon sarcoglycan Epsilon SG Epsilon-sarcoglycan epsilon-sarcoglycan {ECO:0000312 epsilon-SG epsilon-SG {ECO:0000250 ESG e-SG RGD:1303201} sarcoglycan sarcoglycan epsilon sarcoglycan epsilon Sgce sgce {ECO:0000312 UniProtKB:O43556} UNQ433/PRO840; dystonia 11, myoclonic; Epsilon-sarcoglycan; epsilon-sarcoglycan {ECO:0000312|EMBL:AAO37579.1}; Epsilon-SG; epsilon-SG {ECO:0000250|UniProtKB:O43556}; sarcoglycan, epsilon; sgce {ECO:0000312|EMBL:AAO37579.1, ECO:0000312|RGD:1303201}
Gene Aliases: DYT11; e-SG; ESG; SGCE; UNQ433/PRO840
UniProt ID: (Human) O43556, (Mouse) O70258, (Rat) Q6YAT4
Entrez Gene ID: (Human) 8910, (Mouse) 20392, (Rat) 432360
Molecular Function: calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.