Abnova
Manufacturer Code:PAB7603
Catalog # 89-133-408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SFRP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SFRP2.
For Research Use Only
Protein Aliases: FRP-2; SARP-1; SARP1; SDF-5; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; Secreted frizzled-related protein 2; sFRP-2; testicular tissue protein Li 170
Gene Aliases: FKSG12; FRP-2; FRP2; SARP1; SDF-5; SFRP2; UNQ361/PRO697
UniProt ID: (Human) Q96HF1
Entrez Gene ID: (Human) 6423
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.