Proteintech
Manufacturer Code:111472AP150UL
Catalog # 50-172-6916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SEC61G Fusion Protein Ag1624 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: protein transport protein SEC61 gamma subunit; Protein transport protein Sec61 subunit gamma; Sec61 gamma subunit; Sec61 gamma subunit SEC61G SSS1
Gene Aliases: SEC61G; SSS1
UniProt ID: (Human) P60059, (Mouse) P60060
Entrez Gene ID: (Human) 23480, (Rat) 689134, (Mouse) 20335
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.