Invitrogen
Manufacturer Code:PA5112705
Catalog # PIPA5112705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Adseverin protein (556-703AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adseverin; Adseverin AW545522 Gelsolin-like protein KIAA1905 SCIN Scind Scinderin; Gelsolin-like protein; Scinderin
Gene Aliases: adseverin; AW545522; KIAA1905; SCIN
UniProt ID: (Human) Q9Y6U3, (Mouse) Q60604
Entrez Gene ID: (Human) 85477, (Mouse) 20259
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.