Invitrogen
Manufacturer Code:PA5116466
Catalog # PIPA5116466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within Zebrafish SARS2 aa 293-514 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410015F05Rik D7Ertd353e EC 6.1.1.11 mitochondrial seryl-tRNA synthetase mtSerRS SARS Sars2 Sarsm serine tRNA ligase 2 mitochondrial serine-tRNA ligase serine-tRNA ligase mitochondrial serine--tRNA ligase mitochondrial SerRS SerRSmt SERS seryl-aminoacyl-tRNA synthetase 2 Seryl-tRNA synthetase seryl-tRNA synthetase 2 mitochondrial seryl-tRNA synthetase mitochondrial seryl-tRNA(Ser/Sec) synthetase SYS
Gene Aliases: SerRS2; zgc:66345
Entrez Gene ID: (Zebrafish) 394154
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.