Invitrogen
Manufacturer Code:PA5110341
Catalog # PIPA5110341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 289-518 of human SARS2 (NP_060297.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410015F05Rik D7Ertd353e EC 6.1.1.11 mtSerRS SARS SARS2 Sarsm serine-tRNA ligase Serine--tRNA ligase mitochondrial SerRS SerRSmt SERS Seryl-tRNA synthetase Seryl-tRNA(Ser/Sec) synthetase SYS; mitochondrial seryl-tRNA synthetase; serine tRNA ligase 2, mitochondrial; Serine--tRNA ligase, mitochondrial; serine-tRNA ligase, mitochondrial; SerRS; SerRSmt; Seryl-tRNA synthetase; seryl-tRNA synthetase, mitochondrial; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: 2410015F05Rik; D7Ertd353e; mtSerRS; SARS; SARS2; SARSM; SerRS; SerRSmt; SERS; SYS
UniProt ID: (Human) Q9NP81, (Mouse) Q9JJL8
Entrez Gene ID: (Human) 54938, (Mouse) 71984
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.