Novus Biologicals
Manufacturer Code:NBP224747R
Catalog # NBP224747R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Ncap11 |
Immunogen | The antibody was developed by immunizing mice with full-length recombinant SARS nucleocapsid protein (Genbank accession no. NP_828858). The specific epitope has not been determined. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: N N protein N structural protein NC Nucleocapsid protein Nucleoprotein Protein N SARS coronavirus N protein SARS coronavirus nucleocapsid protein SARS CoV SARS CoV N protein SARS CoV nucleocapsid protein SARS N protein SARS Nucleoprotein SARSCoV SARSCoV N protein SARSCoV nucleocapsid protein Severe acute respiratory syndrome; serine tRNA ligase 1, cytoplasmic; Serine--tRNA ligase, cytoplasmic; SerRS; Seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: SARS; SARS1; SERRS; SERS
UniProt ID: (Human) P49591
Entrez Gene ID: (Human) 6301
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.