R&D Systems
Manufacturer Code:MAB3820
Catalog # MAB3820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 397919 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Reelin Ala27-Met862 Accession # Q60841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.- Reeler reelin RELN RL RLPRO1598; Reelin
Gene Aliases: ETL7; LIS2; PRO1598; RELN; RL
UniProt ID: (Human) P78509
Entrez Gene ID: (Human) 5649
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.