R&D Systems
Manufacturer Code:MAB61092SP
Catalog # MAB61092SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 732111 |
Immunogen | <EM>E. coli</EM>-derived recombinant human ROR gamma/RORC/NR1F3 Met1-Gln121 Accession # P51449 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC129539 NR1F3 Nuclear receptor ROR gamma Nuclear receptor ROR-gamma Nuclear receptor RZR gamma Nuclear receptor RZR-gamma Nuclear receptor subfamily 1 group F member 3 RAR-Related Orphan Nuclear Receptor Variant 2 RAR-related orphan receptor C RAR-related orphan receptor gamma retinoic acid-binding receptor gamma retinoid-related orphan receptor gamma Retinoid-related orphan receptor-gamma ROR gamma RORC RORG RORGMGC129539 RZRG RZRGRZR-GAMMA TOR; Nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; Retinoid-related orphan receptor-gamma
Gene Aliases: IMD42; NR1F3; RORC; RORG; RZR-GAMMA; RZRG; TOR
UniProt ID: (Human) P51449
Entrez Gene ID: (Human) 6097
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.