Invitrogen
Manufacturer Code:PA5110291
Catalog # PIPA5110291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 206-315 of human RHBDD1 (NP_115652.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4930418P06Rik HSD50 HSD-50 mRHBDD1 MSD50 MSD-50 RGD1306477 RHBDD1 Rhbdl4 Rhomboid domain-containing protein 1 Rhomboid-like protein 4 Rhomboid-related protein 4 rRHBDD1 RRP4; mRHBDD1; Rhomboid domain-containing protein 1; Rhomboid-like protein 4; Rhomboid-related protein 4; rRHBDD1; RRP4
Gene Aliases: 4930418P06Rik; HSD-50; HSD50; mRHBDD1; MSD-50; MSD50; RGD1306477; RHBDD1; RHBDL4; rRHBDD1; RRP4
UniProt ID: (Human) Q8TEB9, (Rat) Q4V8F3, (Mouse) Q8BHC7
Entrez Gene ID: (Human) 84236, (Rat) 316557, (Mouse) 76867
Molecular Function: aspartic protease hydrolase protease transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.