Invitrogen
Manufacturer Code:PA526892
Catalog # PIPA526892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 265-291 amino acids from the central region of human RGR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RGR-opsin; RP44 RGR-opsin RPE retinal G-protein coupled receptor RPE-retinal G protein-coupled receptor; RPE retinal G-protein coupled receptor; RPE-retinal G protein-coupled receptor
Gene Aliases: RGR; RP44
UniProt ID: (Human) P47804
Entrez Gene ID: (Human) 5995
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.