Novus Biologicals
Manufacturer Code:NBP230891
Catalog # NBP230891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LGLVRKPRRPPPPTMDPPSPSRTSQTQPTATSPLTSYRWHTGGGGEKAAGGFRWGRFAGWGRALSHQEPMVSTQPAPRSIFRRVL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAS Protein Activator Like 3 RAS Protein Activator Like-3; RAS protein activator like-3
Gene Aliases: RASAL3
UniProt ID: (Human) Q86YV0
Entrez Gene ID: (Human) 64926
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.