Novus Biologicals
Manufacturer Code:NBP297779AF488
Catalog # NB047649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human RABGGTA fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.5.1.60 Geranylgeranyl transferase type II subunit alpha geranylgeranyl transferase type-2 subunit alpha protein prenyltransferase alpha subunit repeat containing 3 PTAR3 Rab geranylgeranyltransferase subunit alpha Rab geranyl-geranyltransferase subunit alpha Rab geranylgeranyltransferase alpha subunit Rab GG transferase alpha Rab GGTase alpha; Geranylgeranyl transferase type II subunit alpha; Geranylgeranyl transferase type-2 subunit alpha; protein prenyltransferase alpha subunit repeat containing 3; Rab geranyl-geranyltransferase subunit alpha; Rab geranylgeranyltransferase subunit alpha; Rab geranylgeranyltransferase, alpha subunit; Rab GG transferase alpha; Rab GGTase alpha
Gene Aliases: PTAR3; RABGGTA
UniProt ID: (Human) Q92696
Entrez Gene ID: (Human) 5875
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.