Novus Biologicals
Manufacturer Code:NBP29459120UL
Catalog # NB023707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 370-650 of human QSOX2 (NP_859052.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp762A2013 quiescin Q6 sulfhydryl oxidase 2 quiescin Q6-like 1 quiescin Q6-like protein 1; Neuroblastoma-derived sulfhydryl oxidase; quiescin Q6 sulfhydryl oxidase 2; quiescin Q6-like 1; Quiescin Q6-like protein 1; Sulfhydryl oxidase 2; thiol oxidase 2
Gene Aliases: QSCN6L1; QSOX2; SOXN
UniProt ID: (Human) Q6ZRP7
Entrez Gene ID: (Human) 169714
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.