Invitrogen
Manufacturer Code:MA524263
Catalog # PIMA524263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 583603 |
Immunogen | CHO-derived recombit mouse Plexin A2 Met35-Pro1237 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Plex 2; plexin 2; plexin-2; Plexin-A2; Plexin-A2 Semaphorin receptor OCT PLXNA2 KIAA0463 OCT PLXN2 UNQ209/PRO235 PLXNA2; Semaphorin receptor OCT; transmembrane protein OCT
Gene Aliases: 2810428A13Rik; AA589422; AW457381; KIAA0463; mKIAA0463; OCT; PlexA2; PLXN2; PLXNA2; UNQ209/PRO235
UniProt ID: (Human) O75051, (Mouse) P70207
Entrez Gene ID: (Human) 5362, (Mouse) 18845
Molecular Function: kinase protein kinase protein kinase receptor receptor signaling molecule transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.