Invitrogen
Manufacturer Code:PA5106189
Catalog # PIPA5106189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NCoA3 around the phosphorylation site of Ser857. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010305B15Rik Actr AIB1 AIB-1 Amplified in breast cancer 1 protein amplified in breast cancer-1 protein homolog AW321064 bHLHe42 CAGH16 CBP-interacting protein class E basic helix-loop-helix protein 42 CTG26 KAT13B MGC141848 Ncoa3 NCoA-3 Nuclear receptor coactivator 3 OTTHUMP00000031716 p/Cip pCip RAC3 RAC-3 Receptor-associated coactivator 3 RP5-1049G16.3 SRC3 SRC-3 steroid receptor coactivator protein 3 Thyroid hormone receptor activator molecule 1 TNRC14 TNRC16 Tram1 Tram-1; ACTR; AIB-1; Amplified in breast cancer 1 protein; amplified in breast cancer-1 protein homolog; bHLHe42; CBP-interacting protein; Class E basic helix-loop-helix protein 42; NCoA-3; Nuclear receptor coactivator 3; pCIP; RAC-3; Receptor-associated coactivator 3; SRC-3; Steroid receptor coactivator protein 3; Thyroid hormone receptor activator molecule 1; TRAM-1
Gene Aliases: 2010305B15Rik; ACTR; AIB-1; AIB1; AW321064; BHLHE42; CAGH16; CTG26; KAT13B; NCOA3; p/Cip; pCIP; RAC3; SRC-3; SRC3; TNRC14; TNRC16; TRAM-1; TRAM1
UniProt ID: (Human) Q9Y6Q9
Entrez Gene ID: (Human) 8202, (Mouse) 17979
Molecular Function: acetyltransferase transcription factor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.