Invitrogen
Manufacturer Code:PA5105342
Catalog # PIPA5105342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HPRT1 around the phosphorylation site of Ser110. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C81579 HGPRT HGPRTase HPGRT Hprt HPRT 1 HPRT B Hprt1 hprt1 protein hypoxanthine guanine phosphoribosyl transferase hypoxanthine guanine phosphoribosyl transferase 1 hypoxanthine phosphoribosyl transferase hypoxanthine phosphoribosyltransferase 1 hypoxanthine-guanine phosphoribosyltransferase hypoxanthine-guanine phosphoribosyltransferase 1 id:ibd1344 id:ibd5108 testicular tissue protein Li 89 wu:fc10g09 zgc:56221 zgc:86608; HGPRT; HGPRTase; HPRT B; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; Hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; testicular tissue protein Li 89
Gene Aliases: C81579; HGPRT; Hgprtase; HPGRT; HPRT; HPRT1
UniProt ID: (Human) P00492, (Mouse) P00493, (Rat) P27605
Entrez Gene ID: (Human) 3251, (Mouse) 15452, (Rat) 24465
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.