Invitrogen
Manufacturer Code:PA538112
Catalog # PIPA538112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human DYN1 around the phosphorylation site of Ser774 (R-R-SP-P-T) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-dynamin; B-dynamin D100 DNM DNM1 Dynamin BREDNM19 Dynamin brain EC 3.6.5.5 dynamin-1; D100; Dynamin, brain; Dynamin-1
Gene Aliases: AI838169; DNM; DNM1; EIEE31; Ftfl; Kiaa4093; mKIAA4093
UniProt ID: (Human) Q05193, (Mouse) P39053, (Rat) P21575
Entrez Gene ID: (Human) 1759, (Mouse) 13429, (Rat) 140694
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.