Invitrogen
Manufacturer Code:PA599323
Catalog # PIPA599323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CtBP1 around the phosphorylation site of Ser422. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa BFA-dependent ADP-ribosylation substrate; 50 kDa BFA-dependent ADP-ribosylation substrate 50-kDa BFA-induced ADP-ribosylated substrate 50-kDaBFA-inducedADP-ribosylatedsubstrate BARS BARS-50 brefeldin A-ADP-riboslyated substrate brefeldin A-ribosylated substrate CTBP Ctbp1 CtBP1-L CtBP1-S CtBP3 CtBP3/BARS C-terminal binding protein 1 C-terminal-binding protein 1 C-terminal-binding protein 3 C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate D4S115h D5H4S115 D5H4S115E MGC104684; 50-kDa BFA-induced ADP-ribosylated substrate; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; CtBP1; CtBP3
Gene Aliases: 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; CTBP; CTBP1; CtBP1-L; CtBP1-S; Ctbp3; CtBP3/BARS; D4S115h; D5H4S115; D5H4S115E
UniProt ID: (Human) Q13363, (Rat) Q9Z2F5, (Mouse) O88712
Entrez Gene ID: (Human) 1487, (Rat) 29382, (Mouse) 13016
Molecular Function: dehydrogenase oxidoreductase transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.