Invitrogen
Manufacturer Code:PA517542
Catalog # PIPA517542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding pSer485 of human AMPKa1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-AMP-activated protein kinase catalytic subunit alpha-1 5&prime-AMP-activated protein kinase catalytic alpha-1 chain AMP -activate kinase alpha 1 subunit AMP-activated protein kinase catalytic alpha-1 AMPK alpha 1 AMPK subunit alpha-1 pAMPK; 5'-AMP-activated protein kinase alpha-1 catalytic subunit; 5'-AMP-activated protein kinase catalytic subunit alpha-1; 5'-AMP-activated protein kinase, catalytic alpha-1 chain; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK alpha 1; AMPK alpha-1 chain; AMPK subunit alpha-1; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; protein kinase; protein kinase, AMP-activated, alpha 1 catalytic subunit; Tau-protein kinase PRKAA1
Gene Aliases: AI194361; AI450832; AL024255; AMPK; AMPK1; AMPKa1; AMPKalpha1; C130083N04Rik; PRKAA1
UniProt ID: (Human) Q13131, (Rat) P54645, (Mouse) Q5EG47
Entrez Gene ID: (Chicken) 427185, (Human) 5562, (Rat) 65248, (Mouse) 105787
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.