Novus Biologicals
Manufacturer Code:NBP294134100UL
Catalog # NB022825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PFAS (NP_036525.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGAM synthase; FGAM synthase FGAMS FGAR amidotransferase formylglycinamide ribotide synthetase KIAA0361 phosphoribosylformylglycinamidine synthase PURL; FGAR amidotransferase; Formylglycinamide ribonucleotide amidotransferase; Formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Phosphoribosylformylglycinamidine synthase
Gene Aliases: FGAMS; FGAR-AT; FGARAT; KIAA0361; PFAS; PURL
UniProt ID: (Human) O15067
Entrez Gene ID: (Human) 5198
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.