Novus Biologicals
Manufacturer Code:NBP289399F
Catalog # NB107326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human PTP1B/PTPN1 (NP_002818.1 Glu 2-Asn 321). |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.48 protein tyrosine phosphatase 1B protein tyrosine phosphatase non-receptor type 1 Protein-tyrosine phosphatase 1B PTP-1B PTP1Bprotein tyrosine phosphatase placental tyrosine-protein phosphatase non-receptor type 1; protein tyrosine phosphatase, placental; Protein-tyrosine phosphatase 1B; PTP-1B; Tyrosine-protein phosphatase non-receptor type 1
Gene Aliases: PTP1B; PTPN1
UniProt ID: (Human) P18031
Entrez Gene ID: (Human) 5770
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.