R&D Systems
Manufacturer Code:AF4499SP
Catalog # AF4499SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PTK7/CCK4 Ala31-Ser199 Accession # Q13308 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CCK-4; CCK4 CCK-4 CCK4Colon carcinoma kinase 4 colon carcinoma kinase-4 Protein-tyrosine kinase 7 Pseudo tyrosine kinase receptor 7 PTK7 protein tyrosine kinase 7 Tyrosine-protein kinase-like 7; Colon carcinoma kinase 4; Inactive tyrosine-protein kinase 7; Protein-tyrosine kinase 7; Pseudo tyrosine kinase receptor 7; PTK7 protein tyrosine kinase 7; Tyrosine-protein kinase-like 7
Gene Aliases: CCK-4; CCK4; PTK7
UniProt ID: (Human) Q13308
Entrez Gene ID: (Human) 5754
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase protein kinase receptor receptor transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.