Invitrogen
Manufacturer Code:PA5103514
Catalog # PIPA5103514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PSMD9 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500011J20Rik 2610202L11Rik 26S proteasome non-ATPase regulatory subunit 9 26S proteasome regulatory subunit p27 Bridge Bridge-1 homolog of rat Bridge 1 MGC8644 p27 proteasome (prosome macropain) 26S subunit non-ATPase 9 proteasome 26S non-ATPase regulatory subunit 9 proteasome 26S subunit non-ATPase 9 Psmd9 Rpn4 Transactivating protein Bridge-1; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; homolog of rat Bridge 1; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; Transactivating protein Bridge-1
Gene Aliases: 1500011J20Rik; 2610202L11Rik; Bridge; Bridge-1; P27; PSMD9; Rpn4
UniProt ID: (Human) O00233, (Mouse) Q9CR00, (Rat) Q9WTV5
Entrez Gene ID: (Human) 5715, (Mouse) 67151, (Rat) 161475
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.