Novus Biologicals
Manufacturer Code:NBP133410
Catalog # NBP133410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human PRPS1L1 Recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.6.1 Phosphoribosyl pyrophosphate synthase III phosphoribosyl pyrophosphate synthetase 1-like 1 phosphoribosylpyrophosphate synthetase subunit III PRPS1 PRPS1-like 1 PRPS3Phosphoribosyl pyrophosphate synthase 1-like 1 PRPSL PRS-III ribose-phosphate diphosphokinase catalytic chain III ribose-phosphate pyrophosphokinase 3 ribose-phosphate pyrophosphokinase III; Phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosylpyrophosphate synthetase subunit III; PRPS1-like 1; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; Ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III
Gene Aliases: PRPS1; PRPS1L1; PRPS3; PRPSL; PRS-III
UniProt ID: (Human) P21108
Entrez Gene ID: (Human) 221823
Molecular Function: carbohydrate kinase kinase ligase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.