Invitrogen
Manufacturer Code:PA541794
Catalog # PIPA541794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PRPF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPrp4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4 pre-mRNA processing factor 4 homolog (yeast) HPRP4 HPRP4P PRP4 Prp4p; PRP4/STK/WD splicing factor; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4
Gene Aliases: 1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; mg:ab03a02; PRP4; Prp4p; PRPF4; RP70; SNRNP60; zgc:65943
UniProt ID: (Bovine) Q3MHE2, (Human) O43172, (Mouse) Q9DAW6
Entrez Gene ID: (Bovine) 508735, (Rabbit) 100351462, (Horse) 100050303, (Dog) 474807, (Human) 9128, (Guinea pig) 100733009, (Mouse) 70052, (Rat) 298095, (Zebrafish) 326944
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.