Abnova
Manufacturer Code:MAB4301
Catalog # 89-111-758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 090-18111 |
Immunogen | Native purified human PRL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native PRL.
For Research Use Only
Protein Aliases: decidual prolactin; growth hormone A1; PRL; Prolactin
Gene Aliases: GHA1; PRL
UniProt ID: (Human) P01236
Entrez Gene ID: (Human) 5617
Molecular Function: growth factor peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.