OriGene
Manufacturer Code:TA800501
Catalog # 50-167-8377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 100-306 of human PPT1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ceroid-palmitoyl-palmitoyl-protein thioesterase 1; CLN1 INCL PPT; Palmitoyl-protein hydrolase 1; Palmitoyl-protein thioesterase 1; palmitoyl-protein thioesterase 1 (ceroid-lipofuscinosis, neuronal 1, infantile); PPT-1
Gene Aliases: CLN1; INCL; PPT; PPT1
UniProt ID: (Human) P50897
Entrez Gene ID: (Human) 5538, (Dog) 475316
Molecular Function: esterase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.