Invitrogen
Manufacturer Code:PA544564
Catalog # PIPA544564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human POLB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; DNA-directed DNA polymerase beta; Pol beta; POLB; Polymerase (DNA directed) beta MGC125976; polymerase (DNA directed), beta; polymerase (DNA) beta; polymerase-beta
Gene Aliases: A430088C08Rik; POLB; wu:fa97a05; zgc:109924
UniProt ID: (Human) P06746, (Bovine) Q27958, (Rat) P06766, (Mouse) Q8K409
Entrez Gene ID: (Human) 5423, (Dog) 494001, (Rabbit) 100348691, (Bovine) 614688, (Goat) 102177689, (Horse) 100050102, (Rat) 29240, (Mouse) 18970, (Guinea pig) 100731356, (Zebrafish) 554204
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.