Invitrogen
Manufacturer Code:PA571728
Catalog # PIPA571728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 42-70 amino acids from the N-terminal region of human PNKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain protein 17; MR-1; Myofibrillogenesis regulator 1; Paroxysmal nonkinesiogenic dyskinesia protein; Probable hydrolase PNKD; Probable hydrolase PNKD 3--- Myofibrillogenesis regulator 1 MR-1 Paroxysmal nonkinesiogenic dyskinesia protein Trans-activated by hepatitis C virus core protein 2 PNKD KIAA1184 MR1 TAHCCP2; trans-activated by hepatitis C virus core protein 2
Gene Aliases: 2210013N15Rik; 2810403H05Rik; AI854243; Brp17; Fksg19; Kiaa1184; MNCb-5687; MR-1; Mr1; Pnkd; Tahccp2
UniProt ID: (Mouse) Q69ZP3
Entrez Gene ID: (Mouse) 56695
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.