Abnova
Manufacturer Code:H00005362K
Catalog # 89-210-056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PLXNA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PLXNA2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ11751 FLJ30634 KIAA0463 OCT PLXN2; plexin 2; Plexin-A2; Semaphorin receptor OCT; transmembrane protein OCT
Gene Aliases: KIAA0463; OCT; PLXN2; PLXNA2; UNQ209/PRO235
UniProt ID: (Human) O75051
Entrez Gene ID: (Human) 5362
Molecular Function: kinase protein kinase protein kinase receptor receptor signaling molecule transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.