Abnova
Manufacturer Code:H00005327K
Catalog # 89-210-050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PLAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PLAT peptide using ARM Technology.
For Research Use Only
Protein Aliases: Alteplase; DKFZp686I03148 T-PA TPA; plasminogen/activator kringle; Reteplase; t-PA; t-plasminogen activator; Tissue-type plasminogen activator; Tissue-type plasminogen activator chain A; Tissue-type plasminogen activator chain B
Gene Aliases: PLAT; T-PA; TPA
UniProt ID: (Human) P00750
Entrez Gene ID: (Human) 5327
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.