Invitrogen
Manufacturer Code:MA520597
Catalog # PIMA520597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A3 |
Immunogen | PKMYT1 (NP&lowbar004194.3 400 a.a. &bsim 499 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase Myt1 kinase PKMYT1 MYT1; Myt1 kinase; protein phosphatase 1, regulatory subunit 126
Gene Aliases: MYT1; PKMYT1; PPP1R126
UniProt ID: (Human) Q99640
Entrez Gene ID: (Human) 9088
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.