Invitrogen
Manufacturer Code:PA589199
Catalog # PIPA589199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 231-500 of human PIP5K1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase alpha; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1 beta; phosphatidylinositol-4-phosphate 5-kinase, type I, alpha; PI4P5KIbeta; PIP5K1-alpha; PIP5KIalpha; ptdIns(4)P-5-kinase 1 alpha
Gene Aliases: PI4P5K-I[a]; PIP5K1-alpha; PIP5K1A; Pip5k1b; PIP5KIalpha; Pipk5a
UniProt ID: (Human) Q99755, (Mouse) P70182
Entrez Gene ID: (Human) 8394, (Mouse) 18720, (Rat) 365865
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.