Abnova
Manufacturer Code:H00008394A01
Catalog # 89-105-843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 258 a.a. &bsim 347 a.a) partial recombinant protein with GST tag.,PIP5K1A (NP_003548 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PIP5K1A.
For Research Use Only
Protein Aliases: 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase alpha; 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase type-1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type I, alpha; PIP5K1-alpha; PIP5KIalpha; ptdIns(4)P-5-kinase 1 alpha
Gene Aliases: PIP5K1A
UniProt ID: (Human) Q99755
Entrez Gene ID: (Human) 8394
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.