Invitrogen
Manufacturer Code:PA572180
Catalog # PIPA572180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 23-57amino acids from the N-terminal region of human PIP4K2C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Phosphatidylinositol 5-phosphate 4-kinase type II gamma; Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-4-phosphate 5-kinase, type II, gamma; phosphatidylinositol-5-phosphate 4-kinase type-2 gamma; PI(5)P 4-kinase type II gamma; PIP4KII-gamma; PIP5K2C PI(5)P 4-kinase type II gamma PIP4KII-gamma phosphatidylinositol 4-phosphate 5-kinase phosphatidylinositol-4-phosphate 5-kinase type II gamma phosphatidylinositol-5-phosphate 4-kinase type-2 gamma
Gene Aliases: Pip4k2c; Pip5k2c
UniProt ID: (Mouse) Q91XU3, (Rat) O88370
Entrez Gene ID: (Mouse) 117150, (Rat) 140607
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.