Novus Biologicals
Manufacturer Code:NB100493SS
Catalog # NB100493SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PINK1 antibody was developed using an N-terminal region synthetic peptide made to the human PINK1 protein sequence (between residues 1-50). [UniProt# Q9BXM7] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRPK; BRPK EC 2.7.11.1 FLJ27236 PARK6 Parkinson disease (autosomal recessive) 6 protein kinase BRPK PTEN Induced Kinase 1 PTEN induced putative kinase 1 PTEN-induced putative kinase protein 1 serine/threonine-protein kinase PINK1 mitochondrial; protein kinase BRPK; PTEN-induced putative kinase protein 1; Serine/threonine-protein kinase PINK1, mitochondrial
Gene Aliases: BRPK; PARK6; PINK1
UniProt ID: (Human) Q9BXM7
Entrez Gene ID: (Human) 65018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.