Novus Biologicals
Manufacturer Code:NBP219830
Catalog # NBP219830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PI3 kinase p101. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F730038I15Rik FOAP-2 p101 P101-PI3K Phosphatidylinositol-45-bisphosphate 3-kinase regulatory subunit phosphoinositide-3-kinase regulatory subunit 5 phosphoinositide-3-kinase regulatory subunit polypeptide p101 PI3-kinase p101 subunit PI3-kinase regulatory subunit 5 PtdIns-3-kinase regulatory subunit regulatory subunit 5 p101; p101-PI3K; Phosphatidylinositol 4,5-bisphosphate 3-kinase regulatory subunit; phosphatidylinositol-4,5-bisphosphate 3-kinase regulatory subunit; Phosphoinositide 3-kinase regulatory subunit 5; phosphoinositide-3-kinase, regulatory subunit 5; PI3-kinase p101 subunit; PI3-kinase regulatory subunit 5; Protein FOAP-2; PtdIns-3-kinase p101; PtdIns-3-kinase regulatory subunit
Gene Aliases: F730038I15Rik; FOAP-2; p101; P101-PI3K; PIK3R5
UniProt ID: (Human) Q8WYR1
Entrez Gene ID: (Human) 23533
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.