Invitrogen
Manufacturer Code:PA586409
Catalog # PIPA586409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 230-286 of Human PIG3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p53-induced gene 3 protein; quinone oxidoreductase homolog; Quinone oxidoreductase PIG3; Quinone oxidoreductase PIG3 Tumor protein p53-inducible protein 3 p53-induced gene 3 protein TP53I3 PIG3; Quinone oxidoreductase-like protein 2; Tumor protein p53-inducible protein 3
Gene Aliases: PIG3; RGD1304982; TP53I3
UniProt ID: (Human) Q53FA7, (Rat) B0BNC9
Entrez Gene ID: (Human) 9540, (Rat) 289138
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.