Invitrogen
Manufacturer Code:PA521751
Catalog # PIPA521751
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 342 and 403 of PEX13 (Uniprot IDNo.Q92968) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NALD ZWS peroxin-13 peroxisomal membrane protein PEX13 peroxisome biogenesis factor 13; Peroxin-13; Peroxisomal membrane protein PEX13
Gene Aliases: NALD; PBD11A; PBD11B; PEX13; ZWS
UniProt ID: (Human) Q92968
Entrez Gene ID: (Human) 5194
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.