Abnova
Manufacturer Code:PAB15341
Catalog # 89-117-571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S293 human PDHA1. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDHA1.
For Research Use Only
Protein Aliases: PDHA; PDHCE1A; PDHE1-A type I; PHE1A; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; Pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit
Gene Aliases: PDHA; PDHA1; PDHAD; PDHCE1A; PHE1A
UniProt ID: (Human) P08559
Entrez Gene ID: (Human) 5160
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.