Abnova
Manufacturer Code:PAB16437
Catalog # 89-118-310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human PDE1C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PDE1C.
For Research Use Only
Protein Aliases: 3',5'-cyclic-AMP phosphodiesterase; 3',5'-cyclic-GMP phosphodiesterase; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1C; Cam-PDE 1C; Hcam3; Human 3',5' cyclic nucleotide phosphodiesterase (HSPDE1C1A); phosphodiesterase 1C, calmodulin-dependent 70kDa
Gene Aliases: cam-PDE 1C; hCam-3; Hcam3; PDE1C
UniProt ID: (Human) Q14123
Entrez Gene ID: (Human) 5137
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.