Novus Biologicals
Manufacturer Code:NBP273277IR
Catalog # NBP273277IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7D5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 259-545 of human PDE1A (NP_005010) produced in E.coli. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 3&prime-5&prime cyclic nucleotide phosphodiesterase 61 kDa Cam-PDE calcium/calmodulin-dependent 3&prime-5&prime-cyclic nucleotide phosphodiesterase 1A calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase calmodulin-dependent phosphodiesterase Cam-PDE 1A EC 3.1.4 EC 3.1.4.17 hCam-1 HCAM1 HSPDE1A MGC26303 phosphodiesterase 1A calmodulin-dependent phosphodiesterase-1A; 61 kDa Cam-PDE; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; Cam-PDE 1A; hCam-1; phosphodiesterase 1A, calmodulin-dependent
Gene Aliases: CAM-PDE-1A; HCAM-1; HCAM1; HSPDE1A; PDE1A
UniProt ID: (Human) P54750
Entrez Gene ID: (Human) 5136
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.