Proteintech
Manufacturer Code:147051AP150UL
Catalog # 50-172-9657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PCP4 Fusion Protein Ag6388 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain specific antigen PCP 4 PCP4 PEP 19 PEP19 Purkinje cell protein 4; brain specific polypeptide PEP19; Brain-specific antigen PCP-4; Brain-specific polypeptide PEP-19; Calmodulin regulator protein PCP4; Purkinje cell protein 4
Gene Aliases: P16Rimb19; Pcp-4; PCP4; PEP-19; PEP19; PEPZ19
UniProt ID: (Human) P48539, (Mouse) P63054, (Rat) P63055
Entrez Gene ID: (Human) 5121, (Mouse) 18546, (Rat) 25510
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.